Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2B SIRNA Assays ›
Gene Symbol:
RABL2B
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2B
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
-
Chromosome Location:
Chr.22: 50767492 - 50783705 on Build GRCh38
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.584862
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
120941
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003789.1
NP_001003789.1
4
420
NM_001130919.1
NP_001124391.1
4
416
NM_001130920.1
NP_001124392.1
5
575
NM_001130921.1
NP_001124393.1
5
668
NM_001130922.1
NP_001124394.1
4
420
NM_001130923.1
NP_001124395.1
5
575
NM_007081.2
NP_009012.1
4
416
XM_005261899.2
XP_005261956.1
5
672
XM_011530668.1
XP_011528970.1
5
672
XM_011530669.2
XP_011528971.1
4
419
XM_011530670.1
XP_011528972.1
4
436
XM_011530671.1
XP_011528973.1
5
546
XM_011530673.1
XP_011528975.1
5
672
XM_011530675.1
XP_011528977.1
5
672
XM_017028543.1
XP_016884032.1
5
862
XM_017028544.1
XP_016884033.1
5
767
XM_017028545.1
XP_016884034.1
5
708
XM_017028546.1
XP_016884035.1
5
710
XM_017028547.1
XP_016884036.1
5
862
XM_017028548.1
XP_016884037.1
4
419
XM_017028549.1
XP_016884038.1
5
708
XM_017028550.1
XP_016884039.1
5
546
XM_017028551.1
XP_016884040.1
3
593
XM_017028552.1
XP_016884041.1
5
862
XM_017028553.1
XP_016884042.1
5
764
XM_017028554.1
XP_016884043.1
5
708
XM_017028555.1
XP_016884044.1
5
710
XM_017028556.1
XP_016884045.1
5
862
XM_017028557.1
XP_016884046.1
5
708
XM_017028558.1
XP_016884047.1
5
546
XM_017028559.1
XP_016884048.1
3
593
GenBank mRNA
AF095352.1
-
4
357
AK308268.1
-
4
393
AK313442.1
-
5
672
BC014879.2
-
4
382
3906749
BC024281.1
-
5
618
4580599
BG177165.1
-
416
BT020037.1
-
3
204
BX421088.2
-
5
521
BX432293.2
-
4
362
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
CN342140.1
-
3
261
DB451301.2
-
4
384
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0