Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000078.2 | NP_000069.2 | 11 | 1124 | ||
NM_001286085.1 | NP_001273014.1 | 10 | 944 | |||
GenBank mRNA | AK291586.1 | - | 11 | 1084 | ||
AK297699.1 | - | 10 | 948 | |||
BC025739.1 | - | 11 | 1083 | 5223614 | ||
BT020064.1 | - | 10 | 887 | |||
KU177972.1 | - | 11 | 1067 | |||
KU177973.1 | - | 10 | 887 | |||
M30185.1 | - | 11 | 1197 | |||
M83573.1 | - | 10 | 864 |