Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128204.1 | NP_001121676.1 | 4 | 660 | ||
NM_001128205.1 | NP_001121677.1 | 5 | 828 | |||
NM_001128206.1 | NP_001121678.1 | 4 | 666 | |||
NM_015170.2 | NP_055985.2 | 5 | 822 | |||
XM_006716438.2 | XP_006716501.1 | 5 | 860 | |||
XM_006716439.2 | XP_006716502.1 | 4 | 666 | |||
XM_006716440.3 | XP_006716503.1 | 5 | 822 | |||
XM_011517494.1 | XP_011515796.1 | 4 | 571 | |||
XM_011517495.1 | XP_011515797.1 | 5 | 860 | |||
XM_017013250.1 | XP_016868739.1 | 5 | 860 | |||
GenBank mRNA | AF545571.1 | - | 5 | 811 | ||
AK055607.1 | - | 4 | 612 | |||
AK095341.1 | - | 4 | 660 | |||
AK307951.1 | - | 4 | 439 | |||
AY101175.1 | - | 4 | 613 | |||
BX571746.1 | - | 5 | 773 |