Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 4, 5 | 726 | ||
NM_001319652.1 | NP_001306581.1 | 7, 8 | 851 | |||
NM_001319653.1 | NP_001306582.1 | 5, 6 | 763 | |||
NM_001319655.1 | NP_001306584.1 | 2, 3 | 657 | |||
NM_020841.4 | NP_065892.1 | 5, 6 | 772 | |||
XM_005268621.4 | XP_005268678.1 | 8, 9 | 870 | |||
XM_006719224.2 | XP_006719287.1 | 4, 5 | 731 | |||
XM_011537855.2 | XP_011536157.1 | 5, 6 | 389 | |||
XM_011537856.2 | XP_011536158.1 | 6, 7 | 807 | |||
XM_011537857.2 | XP_011536159.1 | 8, 9 | 861 | |||
XM_017018768.1 | XP_016874257.1 | 5, 6 | 404 | |||
XM_017018769.1 | XP_016874258.1 | 6, 7 | 775 | |||
GenBank mRNA | AB040884.3 | - | 5, 6 | 443 | ||
AF323730.1 | - | 4, 5 | 726 | |||
AF392452.1 | - | 5, 6 | 773 | |||
AK055835.1 | - | 4, 5 | 324 | |||
AK092160.1 | - | 2, 3 | 657 | |||
AK289997.1 | - | 5, 6 | 492 | |||
BC082235.1 | - | 5, 6 | 711 | |||
BC093834.1 | - | 4, 5 | 371 | 7939679 | ||
BC101529.1 | - | 4, 5 | 371 | 8069035 | ||
BC111728.1 | - | 5, 6 | 666 | 40029118 | ||
BX647211.1 | - | 4, 5 | 325 | |||
CR749542.1 | - | 7, 8 | 577 |