Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121113
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
11, 12
1011
NM_148904.3
NP_683702.1
7, 8
728
NM_148905.3
NP_683703.1
6, 7
632
NM_148906.2
NP_683704.2
10, 11
960
NM_148907.2
NP_683705.1
7, 8
605
NM_148908.3
NP_683706.3
10, 11
972
NM_148909.3
NP_683707.3
11, 12
1041
NR_036662.1
10, 11
932
XM_006710318.2
XP_006710381.1
10, 11
771
XM_006710320.3
XP_006710383.1
8, 9
608
XM_006710321.2
XP_006710384.1
7, 8
604
XM_006710322.3
XP_006710385.1
8, 9
765
XM_006710323.2
XP_006710386.1
7, 8
673
XM_006710324.3
XP_006710387.1
9, 10
683
XM_006710325.3
XP_006710388.1
8, 9
653
XM_006710326.2
XP_006710389.1
6, 7
655
XM_011540599.2
XP_011538901.1
12, 13
865
XM_011540600.2
XP_011538902.1
11, 12
832
XM_011540601.2
XP_011538903.1
10, 11
793
XM_011540602.2
XP_011538904.1
11, 12
767
XM_011540603.2
XP_011538905.1
11, 12
791
XM_011540604.2
XP_011538906.1
8, 9
566
XM_017000218.1
XP_016855707.1
9, 10
732
XM_017000219.1
XP_016855708.1
10, 11
752
XM_017000220.1
XP_016855709.1
7, 8
493
XM_017000221.1
XP_016855710.1
7, 8
444
XM_017000222.1
XP_016855711.1
11, 12
977
XM_017000223.1
XP_016855712.1
11, 12
975
XM_017000224.1
XP_016855713.1
8, 9
920
XM_017000225.1
XP_016855714.1
9, 10
771
GenBank mRNA
AF392445.1
-
11, 12
785
AK022554.1
-
6, 7
412
AK027535.1
-
10, 11
742
AK027707.1
-
7, 8
568
AK056617.1
-
11, 12
830
AK091703.1
-
10, 11
714
AK124037.1
-
2, 3
1258
AK128043.1
-
13, 14
1235
AK297656.1
-
8, 9
758
AK315167.1
-
7, 8
604
AY178997.1
-
10, 11
734
BC025978.2
-
7, 8
638
3621193
CR457288.1
-
4, 5
279
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001044234, FQ228495.This siRNA also targets the following transcripts in Mouse: NM_133885, NM_173350, NM_001134791, AK172010, AK215553, and others.
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0