Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 57 | 7053 | ||
XM_005270690.3 | XP_005270747.1 | 44 | 5094 | |||
XM_017000831.1 | XP_016856320.1 | 58 | 7118 | |||
XM_017000832.1 | XP_016856321.1 | 58 | 7115 | |||
XM_017000833.1 | XP_016856322.1 | 58 | 6610 | |||
XM_017000834.1 | XP_016856323.1 | 57 | 6550 | |||
GenBank mRNA | AB028980.1 | - | 16 | 1872 | ||
AK000321.1 | - | 1 | 37 | |||
AL832370.1 | - | 19 | 2279 | |||
BC029660.1 | - | 2 | 106 |