Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206650.1 | NP_001193579.1 | 5 | 1205 | ||
NM_001290042.1 | NP_001276971.1 | 6 | 1571 | |||
NM_002783.2 | NP_002774.2 | 6 | 1571 | |||
GenBank mRNA | BC030979.1 | - | 6 | 1566 | ||
BC136401.1 | - | 5 | 1125 | |||
U18467.1 | - | 6 | 1521 |