Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145364.2 | NP_001138836.1 | 2 | 326 | ||
NM_001301058.1 | NP_001287987.1 | 2 | 326 | |||
NM_001321382.1 | NP_001308311.1 | 2 | 326 | |||
NM_001321383.1 | NP_001308312.1 | 2 | 326 | |||
NM_148912.3 | NP_683710.1 | 2 | 326 | |||
NM_148913.3 | NP_683711.1 | 2 | 305 | |||
NR_026912.2 | 2 | 326 | ||||
NR_135627.1 | 2 | 326 | ||||
XM_006716144.2 | XP_006716207.1 | 1 | 24 | |||
XM_011516590.2 | XP_011514892.1 | 2 | 290 | |||
XM_011516591.1 | XP_011514893.1 | 2 | 226 | |||
GenBank mRNA | AF217971.1 | - | 2 | 240 | ||
AF412030.1 | - | 2 | 318 | |||
AF412031.1 | - | 2 | 318 | |||
AF412032.1 | - | 2 | 318 | |||
AY053499.1 | - | 2 | 297 | |||
AY053500.1 | - | 2 | 297 | |||
BC008251.1 | - | 2 | 290 | 3873594 | ||
BC011712.2 | - | 2 | 244 | |||
BC067750.1 | - | 2 | 261 | 5265402 | ||
BG722364.1 | - | 2 | 318 | |||
BM557977.1 | - | 2 | 326 | |||
BU175566.1 | - | 2 | 209 | |||
BU189995.1 | - | 2 | 282 | |||
CB998040.1 | - | 2 | 318 | |||
DB460434.1 | - | 2 | 319 | |||
DR006453.1 | - | 2 | 295 |