Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004825.2 | NP_004816.1 | 1 | 788 | ||
GenBank mRNA | AF080598.1 | - | 1 | 788 | ||
AK292233.1 | - | 1 | 761 |
Gene Symbol: CDY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001722.1 | NP_001001722.1 | 1 | 787 | ||
GenBank mRNA | BC069087.1 | - | 1 | 787 | 7216901 | |
BC130426.1 | - | 1 | 556 | 40146457 | ||
BC130428.1 | - | 1 | 556 | 40146459 |