Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167.3 | NP_001158.2 | 7 | 3305 | ||
NM_001204401.1 | NP_001191330.1 | 7 | 3272 | |||
NR_037916.1 | 6 | 2396 | ||||
XM_006724754.2 | XP_006724817.1 | 8 | 3385 |