Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001375.2 | NP_001366.1 | 6 | 1218 | ||
GenBank mRNA | AB004574.1 | - | 6 | 1144 | ||
AF045937.1 | - | 6 | 1148 | |||
AF047016.1 | - | 6 | 1144 | |||
AF060222.1 | - | 6 | 1165 | |||
AK075449.1 | - | 6 | 1133 | |||
AK297493.1 | - | 5 | 981 | |||
AK311145.1 | - | 6 | 1142 | |||
AK315358.1 | - | 6 | 1160 | |||
BC010419.2 | - | 6 | 1096 | 3528108 | ||
BC065209.1 | - | 6 | 1163 | 5502061 | ||
BT007047.1 | - | 6 | 1072 | |||
FJ752529.1 | - | 6 | 1024 |