Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003012.4 | NP_003003.3 | 3 | 2943 | ||
GenBank mRNA | AF056087.1 | - | 3 | 2926 | ||
AK127331.1 | - | 3 | 2355 | |||
BC036503.1 | - | 3 | 2949 | 5265277 | ||
DA571541.1 | - | 1 | 456 |
Set Membership: |
G-protein coupled receptor |