Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121426
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
7
1045
NM_001243226.2
NP_001230155.2
8
835
NM_001243227.1
NP_001230156.1
6
879
NM_001243228.1
NP_001230157.1
7
971
NM_001243230.1
NP_001230159.1
6
521
NM_001243231.1
NP_001230160.1
5
479
NM_001243232.1
NP_001230161.1
3
501
NM_001243233.1
NP_001230162.1
3
442
NM_001306207.1
NP_001293136.1
6
670
NM_001306208.1
NP_001293137.1
3
501
NM_003199.2
NP_003190.1
7
1045
XM_005266739.3
XP_005266796.2
6
685
XM_005266741.3
XP_005266798.1
7
645
XM_005266743.4
XP_005266800.1
6
506
XM_005266744.3
XP_005266801.1
6
626
XM_005266745.3
XP_005266802.1
6
488
XM_005266747.3
XP_005266804.1
5
482
XM_005266749.3
XP_005266806.1
3
273
XM_005266752.4
XP_005266809.1
3
843
XM_005266754.3
XP_005266811.1
2
267
XM_005266755.4
XP_005266812.1
3
302
XM_006722536.2
XP_006722599.1
7
968
XM_006722537.2
XP_006722600.1
7
601
XM_006722538.2
XP_006722601.1
6
623
XM_011526158.2
XP_011524460.1
6
1599
XM_011526160.2
XP_011524462.1
2
652
XM_017025934.1
XP_016881423.1
6
682
XM_017025935.1
XP_016881424.1
6
685
XM_017025936.1
XP_016881425.1
6
685
XM_017025937.1
XP_016881426.1
6
439
XM_017025938.1
XP_016881427.1
7
598
XM_017025939.1
XP_016881428.1
7
648
XM_017025940.1
XP_016881429.1
7
968
XM_017025941.1
XP_016881430.1
7
601
XM_017025942.1
XP_016881431.1
7
645
XM_017025943.1
XP_016881432.1
7
648
XM_017025944.1
XP_016881433.1
6
903
XM_017025945.1
XP_016881434.1
6
906
XM_017025946.1
XP_016881435.1
6
920
XM_017025947.1
XP_016881436.1
6
506
XM_017025948.1
XP_016881437.1
6
903
XM_017025949.1
XP_016881438.1
6
623
XM_017025950.1
XP_016881439.1
6
903
XM_017025951.1
XP_016881440.1
5
482
XM_017025952.1
XP_016881441.1
3
496
XM_017025953.1
XP_016881442.1
3
273
XM_017025954.1
XP_016881443.1
3
857
XM_017025955.1
XP_016881444.1
2
591
XM_017025956.1
XP_016881445.1
2
267
GenBank mRNA
AB209741.1
-
2
153
AK095041.1
-
3
501
AK096862.1
-
5
479
AK122765.1
-
7
1045
AK299169.1
-
6
879
AK300612.1
-
3
442
AK300636.1
-
2
172
AK301144.1
-
6
656
AK315074.1
-
7
959
AK316165.1
-
2
216
BC031056.1
-
8
796
BC125084.1
-
6
459
40117795
BC125085.1
-
6
459
40117798
BP293388.1
-
6
508
FR748208.1
-
2
88
FR748209.1
-
2
589
FR748210.1
-
6
433
FR748211.1
-
6
433
FR748214.1
-
5
361
FR748216.1
-
6
427
FR748217.1
-
5
307
FR748218.1
-
1
43
FR748219.1
-
3
220
M74719.1
-
7
632
X52079.1
-
4
298
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0