Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002814.3 | NP_002805.1 | 1 | 201 | ||
NM_170750.2 | NP_736606.1 | 1 | 201 | |||
GenBank mRNA | AB009619.1 | - | 1 | 99 | ||
AK058108.1 | - | 1 | 175 | |||
AK295996.1 | - | 1 | 95 | |||
AK309326.1 | - | 1 | 95 | |||
AY057056.1 | - | 1 | 99 | |||
BC011960.1 | - | 1 | 110 | 3881328 | ||
BI114640.1 | - | 1 | 202 | |||
BQ217943.1 | - | 1 | 117 | |||
BT019689.1 | - | 1 | 77 | |||
D83197.1 | - | 1 | 175 |