Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256574.1 | NP_001243503.1 | 4 | 3997 | ||
NM_001256575.1 | NP_001243504.1 | 4 | 3985 | |||
NM_001256576.1 | NP_001243505.1 | 3 | 3724 | |||
NM_003633.3 | NP_003624.1 | 3 | 3860 | |||
NR_046318.1 | 2 | 2045 | ||||
XM_011543696.2 | XP_011541998.1 | 4 | 4068 | |||
XM_011543697.2 | XP_011541999.1 | 4 | 3914 | |||
GenBank mRNA | AF059611.1 | - | 3 | 3128 | ||
AU120246.1 | - | 1 | 134 |