Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK075140.1 | - | 5, 6 | 491 | ||
AK309540.1 | - | 6, 7 | 520 | |||
AY358081.1 | - | 5, 6 | 480 | |||
AY358283.1 | - | 5, 6 | 517 | |||
BC020731.1 | - | 5, 6 | 491 | 4692272 | ||
CR456808.1 | - | 6, 7 | 518 |