Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256560.1 | NP_001243489.1 | 4 | 504 | ||
NM_004944.3 | NP_004935.1 | 5 | 594 | |||
GenBank mRNA | AB209613.1 | - | 2 | 3120 | ||
AF047354.1 | - | 5 | 567 | |||
AK131323.1 | - | 8 | 1089 | |||
AK301263.1 | - | 4 | 470 | |||
AK313303.1 | - | 5 | 560 | |||
BC015831.1 | - | 5 | 564 | 4723725 | ||
DB164811.1 | - | 2 | 91 | |||
U56814.1 | - | 5 | 521 |