Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002824.4 | NP_002815.3 | 3 | 480 | ||
XM_011520987.1 | XP_011519289.1 | 3 | 793 | |||
GenBank mRNA | BC017025.1 | - | 3 | 199 | 3900998 | |
BC128229.1 | - | 4 | 147 | |||
BM550529.1 | - | 557 | ||||
M24398.1 | - | 3 | 449 |