Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244984.1 | NP_001231913.1 | 5, 6 | 741 | ||
NM_005065.5 | NP_005056.3 | 5, 6 | 741 | |||
XM_005267988.2 | XP_005268045.1 | 5, 6 | 624 | |||
XM_005267989.3 | XP_005268046.1 | 5, 6 | 744 | |||
GenBank mRNA | AB020335.1 | - | 5, 6 | 669 | ||
AF052059.1 | - | 5, 6 | 717 | |||
AK075511.1 | - | 5, 6 | 720 | |||
AY358651.1 | - | 5, 6 | 713 |