Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EXOG SIRNA Assays ›
Gene Symbol:
EXOG
Gene Name:
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases:
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB
Chromosome Location:
Chr.3: 38496127 - 38526305 on Build GRCh38
UniGene ID:
Hs.517897
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121519
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EXOG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145464.1
NP_001138936.1
2
299
NM_001318954.1
NP_001305883.1
4
419
NM_001318955.1
NP_001305884.1
5
517
NM_001318956.1
NP_001305885.1
4
516
NM_001318957.1
NP_001305886.1
6
655
NM_001318958.1
NP_001305887.1
6
573
NM_001318959.1
NP_001305888.1
5
611
NM_005107.3
NP_005098.2
3
449
NR_134938.1
3
355
XM_005265633.3
XP_005265690.1
3
295
XM_005265635.3
XP_005265692.1
3
398
XM_006713442.3
XP_006713505.1
4
681
XM_006713444.3
XP_006713507.1
3
292
XM_017007584.1
XP_016863073.1
3
3610
XM_017007585.1
XP_016863074.1
4
2420
XM_017007586.1
XP_016863075.1
3
2345
XM_017007587.1
XP_016863076.1
5
500
XM_017007588.1
XP_016863077.1
5
745
XM_017007589.1
XP_016863078.1
6
812
XM_017007590.1
XP_016863079.1
4
302
XM_017007591.1
XP_016863080.1
4
299
XM_017007592.1
XP_016863081.1
5
411
XM_017007593.1
XP_016863082.1
4
221
GenBank mRNA
AB020523.1
-
3
380
AK023235.1
-
3
356
AK289602.1
-
6
480
AK290107.1
-
4
419
AK301067.1
-
2
228
AK308814.1
-
3
283
AK315814.1
-
5
386
BC104212.1
-
4
445
40031649
BC104213.1
-
5
448
40031650
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
endonuclease activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0