Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318956.1 | NP_001305885.1 | 2 | 344 | ||
NM_001318957.1 | NP_001305886.1 | 2 | 344 | |||
NM_001318959.1 | NP_001305888.1 | 3 | 431 | |||
NM_005107.3 | NP_005098.2 | 2 | 344 | |||
XM_005265635.3 | XP_005265692.1 | 2 | 293 | |||
XM_006713442.3 | XP_006713505.1 | 3 | 576 | |||
XM_017007584.1 | XP_016863073.1 | 2 | 271 | |||
XM_017007585.1 | XP_016863074.1 | 2 | 271 | |||
XM_017007586.1 | XP_016863075.1 | 2 | 271 | |||
XM_017007587.1 | XP_016863076.1 | 2 | 261 | |||
XM_017007588.1 | XP_016863077.1 | 3 | 576 | |||
XM_017007589.1 | XP_016863078.1 | 3 | 576 | |||
XM_017007590.1 | XP_016863079.1 | 2 | 122 | |||
XM_017007591.1 | XP_016863080.1 | 2 | 130 | |||
GenBank mRNA | AB020523.1 | - | 2 | 275 | ||
AK023235.1 | - | 2 | 251 | |||
BC104212.1 | - | 2 | 273 | 40031649 |