Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 2 | 310 | ||
NM_001024960.2 | NP_001020131.1 | 2 | 165 | |||
NM_001198780.1 | NP_001185709.1 | 2 | 164 | |||
NM_005718.4 | NP_005709.1 | 2 | 676 | |||
GenBank mRNA | AF006087.1 | - | 2 | 117 | ||
AF019888.1 | - | 1 | 102 | |||
AW408422.1 | - | 2 | 123 | |||
BC012596.2 | - | 1 | 99 | |||
BG938423.1 | - | 2 | 148 | |||
BI768875.1 | - | 2 | 289 | |||
BI910730.1 | - | 2 | 684 | |||
BM045872.1 | - | 2 | 175 | |||
BQ688329.1 | - | 2 | 248 | |||
BX419672.2 | - | 2 | 164 | |||
DA226584.1 | - | 2 | 165 | |||
DB060690.1 | - | 2 | 310 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 2 | 676 | ||
GenBank mRNA | AK024110.1 | - | 2 | 666 | ||
BC025289.1 | - | 1 | 110 |