Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 1 | 173 | ||
GenBank mRNA | AK001467.1 | - | 1 | 145 | ||
BC001238.1 | - | 1 | 102 | 3452935 | ||
BC003360.1 | - | 1 | 132 | 2900252 | ||
BC024739.1 | - | 1 | 147 | 4995871 | ||
BU599536.1 | - | 1 | 191 |