Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 10 | 1481 | ||
GenBank mRNA | AB209392.1 | - | 3 | 548 | ||
AK001467.1 | - | 10 | 1453 | |||
AK091227.1 | - | 8 | 908 | |||
BC001238.1 | - | 10 | 1410 | 3452935 | ||
BC003360.1 | - | 10 | 1440 | 2900252 | ||
BC024739.1 | - | 10 | 1455 | 4995871 | ||
CR457060.1 | - | 9 | 1202 | |||
X98743.1 | - | 9 | 1219 |