Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039937.1 | NP_001035026.1 | 19 | 2711 | ||
NM_001306091.1 | NP_001293020.1 | 20 | 3487 | |||
NM_012141.2 | NP_036273.1 | 18 | 3203 | |||
XM_011535040.2 | XP_011533342.1 | 18 | 3300 | |||
XM_011535041.1 | XP_011533343.1 | 17 | 3116 | |||
XM_017020540.1 | XP_016876029.1 | 17 | 2661 | |||
GenBank mRNA | AF097645.1 | - | 18 | 3065 | ||
AK074946.1 | - | 16 | 2293 | |||
AK096696.1 | - | 15 | 2423 | |||
AK128795.1 | - | 16 | 2358 | |||
AK304218.1 | - | 17 | 3005 | |||
AL117626.1 | - | 9 | 1358 | |||
AL833524.1 | - | 19 | 2779 | |||
BC039829.1 | - | 18 | 3077 | 5736353 |