Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287242.1 | NP_001274171.1 | 4 | 644 | ||
NM_006125.2 | NP_006116.2 | 4 | 1867 | |||
NM_013423.2 | NP_038267.1 | 4 | 696 | |||
NM_013427.2 | NP_038286.2 | 4 | 1867 | |||
NR_109776.1 | 5 | 1963 | ||||
GenBank mRNA | AB208792.1 | - | 4 | 1249 | ||
AF012272.2 | - | 4 | 1868 | |||
AF022212.2 | - | 5 | 1964 | |||
AF117067.1 | - | 4 | 1868 | |||
AF177663.1 | - | 4 | 696 | |||
AF177665.1 | - | 4 | 502 | |||
AK293078.1 | - | 4 | 1298 | |||
AK297713.1 | - | 4 | 541 | |||
AK303444.1 | - | 4 | 557 | |||
BC150635.1 | - | 4 | 1026 | 9057005 |