Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TONSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013432.4 | NP_038460.4 | 23 | 3713 | ||
XM_011517048.2 | XP_011515350.1 | 16 | 2812 | |||
XM_011517049.2 | XP_011515351.1 | 16 | 2773 | |||
GenBank mRNA | AB209126.1 | - | 14 | 5800 | ||
AK097406.1 | - | 2 | 1728 | |||
BC008782.2 | - | 22 | 3685 | 3628374 |