Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 5 | 1374 | ||
GenBank mRNA | AF113537.1 | - | 5 | 1241 | ||
AF133422.1 | - | 5 | 1217 | |||
AK098398.1 | - | 1782 | ||||
AK295206.1 | - | 4 | 1057 | |||
AK297196.1 | - | 5 | 1180 | |||
AK315949.1 | - | 3 | 989 | |||
AK315952.1 | - | 4 | 1071 | |||
BC002619.2 | - | 5 | 1257 | 3160345 | ||
BC018873.2 | - | 5 | 1250 | |||
CD627354.1 | - | 1 | 427 |