Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304746.1 | NP_001291675.1 | 3 | 610 | ||
NM_017821.4 | NP_060291.2 | 3 | 640 | |||
GenBank mRNA | AK097621.1 | - | 3 | 1375 | ||
AK294115.1 | - | 3 | 610 | |||
AY126343.1 | - | 2 | 366 | |||
BC013103.1 | - | 3 | 611 | |||
BC137108.1 | - | 2 | 489 | 9021105 | ||
BC137110.1 | - | 2 | 489 | 9021107 | ||
BC144476.1 | - | 2 | 489 |