Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DZANK1 SIRNA Assays ›
Gene Symbol:
DZANK1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2
Chromosome Location:
Chr.20: 18383367 - 18467185 on Build GRCh38
UniGene ID:
Hs.472225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121822
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DZANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099407.1
NP_001092877.1
13
1570
XM_005260742.3
XP_005260799.1
13
1622
XM_005260743.3
XP_005260800.1
12
1476
XM_006723584.2
XP_006723647.1
11
1397
XM_011529268.2
XP_011527570.1
13
1740
XM_011529271.2
XP_011527573.1
13
2353
XM_011529272.2
XP_011527574.1
13
1736
XM_011529275.2
XP_011527577.1
12
2226
XM_011529277.2
XP_011527579.1
5
2467
XM_017027909.1
XP_016883398.1
13
1723
XM_017027910.1
XP_016883399.1
13
1606
XM_017027911.1
XP_016883400.1
13
1602
XM_017027912.1
XP_016883401.1
13
2309
XM_017027913.1
XP_016883402.1
13
2294
XM_017027914.1
XP_016883403.1
13
1548
XM_017027915.1
XP_016883404.1
13
1544
XM_017027916.1
XP_016883405.1
11
2108
XM_017027917.1
XP_016883406.1
14
2403
XM_017027918.1
XP_016883407.1
14
1657
XM_017027919.1
XP_016883408.1
14
1653
XM_017027920.1
XP_016883409.1
12
1612
XM_017027921.1
XP_016883410.1
6
774
GenBank mRNA
AK001462.1
-
7
757
AK055544.1
-
12
1365
AK131451.1
-
14
2209
AK295073.1
-
13
1480
AK299746.1
-
13
1493
BC036231.1
-
13
2818
BC130341.1
-
12
1398
40146372
BC144153.1
-
12
1398
9052673
CR749327.1
-
14
1622
DQ104738.1
-
4
491
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy in response to mitochondrial depolarization
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-8nwch:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0