Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287516.1 | NP_001274445.1 | 9, 10 | 1091 | ||
NM_001287517.1 | NP_001274446.1 | 9, 10 | 1049 | |||
NM_001287519.1 | NP_001274448.1 | 9, 10 | 822 | |||
NM_001287520.1 | NP_001274449.1 | 9, 10 | 864 | |||
NM_001287522.1 | NP_001274451.1 | 8, 9 | 741 | |||
NM_001287524.1 | NP_001274453.1 | 5, 6 | 646 | |||
NM_004358.4 | NP_004349.1 | 9, 10 | 1674 | |||
NM_021872.3 | NP_068658.1 | 8, 9 | 1593 | |||
NM_021873.3 | NP_068659.1 | 9, 10 | 1716 | |||
NR_136335.1 | 9, 10 | 864 | ||||
NR_136336.1 | 9, 10 | 1091 | ||||
GenBank mRNA | AK092713.1 | - | 9, 10 | 864 | ||
AK295573.1 | - | 9, 10 | 933 | |||
AK299192.1 | - | 9, 10 | 867 | |||
BC006395.2 | - | 9, 10 | 997 | 4135465 | ||
BC009953.2 | - | 9, 10 | 1077 | 4130804 | ||
BC051711.1 | - | 9, 10 | 1621 | 5301205 | ||
BX640836.1 | - | 5, 6 | 638 | |||
BX647988.1 | - | 9, 10 | 779 | |||
FR695900.1 | - | 9, 10 | 864 | |||
FR695901.1 | - | 8, 9 | 741 | |||
M81934.1 | - | 9, 10 | 953 | |||
S78187.1 | - | 9, 10 | 1121 | |||
Z68092.1 | - | 8, 9 | 901 |