Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012398.2 | NP_001012398.1 | 7 | 775 | ||
NM_001308325.1 | NP_001295254.1 | 7 | 746 | |||
NM_022476.3 | NP_071921.1 | 7 | 746 | |||
XM_005256095.4 | XP_005256152.1 | 7 | 1095 | |||
XM_005256096.4 | XP_005256153.1 | 7 | 673 | |||
XM_005256097.4 | XP_005256154.1 | 7 | 707 | |||
XM_005256098.4 | XP_005256155.1 | 7 | 671 | |||
XM_017023564.1 | XP_016879053.1 | 7 | 1095 | |||
XM_017023565.1 | XP_016879054.1 | 7 | 673 | |||
XM_017023566.1 | XP_016879055.1 | 7 | 671 | |||
GenBank mRNA | AK023320.1 | - | 7 | 681 | ||
AK222743.1 | - | 7 | 678 | |||
BC001134.2 | - | 7 | 694 | 2967019 | ||
BC095401.1 | - | 7 | 668 | 30334599 | ||
CR457308.1 | - | 6 | 518 |