Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 1, 2 | 331 | ||
NM_024948.3 | NP_079224.1 | 1, 2 | 331 | |||
XM_006717508.2 | XP_006717571.1 | 1, 2 | 308 | |||
XM_011519690.2 | XP_011517992.1 | 1, 2 | 308 | |||
XM_011519691.2 | XP_011517993.1 | 1, 2 | 308 | |||
XM_017016671.1 | XP_016872160.1 | 1, 2 | 308 | |||
XM_017016673.1 | XP_016872162.1 | 1, 2 | 308 | |||
XM_017016674.1 | XP_016872163.1 | 1, 2 | 308 | |||
GenBank mRNA | AB014761.1 | - | 1, 2 | 97 | ||
AF176916.1 | - | 1, 2 | 292 | |||
AK023459.1 | - | 1, 2 | 318 | |||
AL833873.1 | - | 1, 2 | 293 | |||
BC020605.1 | - | 1, 2 | 296 | |||
BC061585.1 | - | 1, 2 | 255 | |||
BC067799.1 | - | 1, 2 | 317 | 5278533 | ||
DA063140.1 | - | 1, 2 | 331 |