Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024948.3 | NP_079224.1 | 5 | 683 | ||
XM_005252600.2 | XP_005252657.1 | 4 | 577 | |||
XM_006717508.2 | XP_006717571.1 | 5 | 660 | |||
XM_011519690.2 | XP_011517992.1 | 5 | 660 | |||
XM_011519691.2 | XP_011517993.1 | 5 | 660 | |||
XM_011519693.1 | XP_011517995.1 | 4 | 577 | |||
XM_011519694.1 | XP_011517996.1 | 6 | 724 | |||
XM_017016671.1 | XP_016872160.1 | 5 | 660 | |||
XM_017016672.1 | XP_016872161.1 | 6 | 724 | |||
XM_017016673.1 | XP_016872162.1 | 5 | 686 | |||
XM_017016675.1 | XP_016872164.1 | 4 | 577 | |||
GenBank mRNA | AB014761.1 | - | 5 | 449 | ||
AK023459.1 | - | 5 | 670 | |||
AL833873.1 | - | 5 | 645 | |||
BC061585.1 | - | 5 | 607 | |||
BC067799.1 | - | 5 | 669 | 5278533 | ||
CR457315.1 | - | 5 | 449 |