Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025227.2 | NP_079503.1 | 2, 3 | 322 | ||
GenBank mRNA | AF465765.1 | - | 1, 2 | 105 | ||
AK027068.1 | - | 2, 3 | 299 | |||
AK172819.1 | - | 2, 3 | 301 | |||
AK304064.1 | - | 2, 3 | 227 | |||
AY358723.1 | - | 2, 3 | 230 | |||
BC034415.1 | - | 2, 3 | 176 | 5173032 | ||
BP333980.1 | - | 2, 3 | 321 |