Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025227.2 | NP_079503.1 | 16 | 1684 | ||
GenBank mRNA | AK027068.1 | - | 16 | 1662 | ||
AK172819.1 | - | 16 | 1663 | |||
AK304064.1 | - | 16 | 1514 | |||
AY358723.1 | - | 16 | 1592 | |||
BC034415.1 | - | 16 | 1538 | 5173032 |