Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030775.2 | NP_110402.2 | 5 | 1949 | ||
NM_032642.2 | NP_116031.1 | 5 | 1987 | |||
GenBank mRNA | AY009399.1 | - | 5 | 1952 | ||
BC001749.1 | - | 5 | 1988 | 3528819 |