Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033341.4 | NP_203127.3 | 1 | 1927 | ||
GenBank mRNA | AF420440.1 | - | 1 | 1926 | ||
BC046168.1 | - | 1 | 1671 | |||
BC071665.1 | - | 1 | 1475 | 5171408 |