Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323005.1 | NP_001309934.1 | 1, 2 | 353 | ||
NM_145315.4 | NP_660358.2 | 1, 2 | 353 | |||
XM_011535657.2 | XP_011533959.1 | 1, 2 | 236 | |||
XM_011535659.2 | XP_011533961.1 | 1, 2 | 236 | |||
XM_011535660.2 | XP_011533962.1 | 1, 2 | 236 | |||
XM_011535661.2 | XP_011533963.1 | 1, 2 | 236 | |||
GenBank mRNA | AF520418.1 | - | 1, 2 | 333 | ||
BC018445.1 | - | 1, 2 | 333 | 3871205 | ||
BP246728.1 | - | 1, 2 | 361 |