Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178127.4 | NP_835228.2 | 5 | 958 | ||
GenBank mRNA | AK291523.1 | - | 4 | 422 | ||
AY169281.1 | - | 4 | 416 | |||
AY358827.1 | - | 4 | 406 | |||
BC049170.1 | - | 5 | 961 | 5285447 | ||
BC053957.1 | - | 5 | 961 |