Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182658.2 | NP_872599.1 | 3 | 463 | ||
GenBank mRNA | AF549189.1 | - | 3 | 370 | ||
BC113398.1 | - | 3 | 463 | 8322450 | ||
BC113400.1 | - | 3 | 463 | 8322452 |