Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PORCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282167.1 | NP_001269096.1 | 4 | 685 | ||
NM_022825.3 | NP_073736.2 | 4 | 540 | |||
NM_203473.2 | NP_982299.1 | 4 | 540 | |||
NM_203474.1 | NP_982300.1 | 3 | 395 | |||
NM_203475.2 | NP_982301.1 | 4 | 540 | |||
XM_005272635.2 | XP_005272692.1 | 4 | 895 | |||
XM_005272636.2 | XP_005272693.1 | 4 | 895 | |||
XM_005272637.1 | XP_005272694.1 | 5 | 822 | |||
XM_006724545.3 | XP_006724608.1 | 5 | 605 | |||
XM_006724546.3 | XP_006724609.1 | 5 | 677 | |||
XM_006724547.1 | XP_006724610.1 | 4 | 397 | |||
XM_011543948.2 | XP_011542250.1 | 4 | 824 | |||
XM_017029735.1 | XP_016885224.1 | 4 | 905 | |||
XM_017029736.1 | XP_016885225.1 | 4 | 397 | |||
GenBank mRNA | AF317058.1 | - | 3 | 361 | ||
AF317059.1 | - | 3 | 361 | |||
AF317060.1 | - | 3 | 361 | |||
AF317061.1 | - | 3 | 361 | |||
AK056340.1 | - | 4 | 854 | |||
AK057727.1 | - | 4 | 397 | |||
AK293347.1 | - | 4 | 650 | |||
AK314745.1 | - | 3 | 410 | |||
AK316378.1 | - | 3 | 432 | |||
AL538245.3 | - | 4 | 877 | |||
AL579208.3 | - | 4 | 459 | |||
BC019080.2 | - | 4 | 487 | 4559533 | ||
BM543485.1 | - | 4 | 511 | |||
BM545893.1 | - | 411 | ||||
L08239.1 | - | 2 | 277 |