Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017573.4 | NP_060043.2 | 15 | 2553 | ||
XM_005259586.1 | XP_005259643.1 | 14 | 2306 | |||
XM_011528085.2 | XP_011526387.1 | 16 | 2778 | |||
XM_011528086.2 | XP_011526388.1 | 16 | 2751 | |||
XM_011528087.2 | XP_011526389.1 | 15 | 2604 | |||
XM_011528088.2 | XP_011526390.1 | 16 | 2430 | |||
XM_011528089.2 | XP_011526391.1 | 14 | 2496 | |||
XM_011528090.2 | XP_011526392.1 | 13 | 2415 | |||
XM_011528091.2 | XP_011526393.1 | 14 | 2251 | |||
XM_011528093.2 | XP_011526395.1 | 15 | 2653 | |||
XM_011528094.1 | XP_011526396.1 | 13 | 2385 | |||
XM_011528096.1 | XP_011526398.1 | 13 | 2113 | |||
GenBank mRNA | AK057235.1 | - | 12 | 2491 | ||
AL043305.1 | - | 1 | 192 | |||
AL133566.1 | - | 3 | 875 | |||
AY358963.1 | - | 15 | 2511 | |||
BC036354.1 | - | 5 | 1253 | |||
BC142942.1 | - | 10 | 2704 |