Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYSND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040273.2 | NP_001035363.1 | 2 | 1598 | ||
NM_173555.3 | NP_775826.2 | 4 | 1915 | |||
NR_073580.1 | 2 | 366 | ||||
NR_073581.1 | 4 | 803 | ||||
NR_073582.1 | 4 | 683 | ||||
NR_073591.1 | 4 | 594 | ||||
NR_073592.1 | 2 | 486 | ||||
NR_073593.1 | 4 | 586 | ||||
NR_073594.1 | 4 | 706 | ||||
NR_073595.1 | 4 | 839 | ||||
GenBank mRNA | AK074968.1 | - | 4 | 741 | ||
AK097697.1 | - | 3 | 1458 | |||
BC016840.1 | - | 4 | 1369 | |||
BC030242.1 | - | 2 | 1514 | 5176098 | ||
BC047424.1 | - | 2 | 366 | |||
BC111501.1 | - | 4 | 1854 | 5732356 | ||
CD671353.1 | - | 4 | 525 | |||
HY013216.1 | - | 2 | 457 |