Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322.2 | NP_001313.1 | 1 | 106 | ||
GenBank mRNA | BC062679.1 | - | 1 | 164 | 30350087 |