Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MDM2 SIRNA Assays ›
Gene Symbol:
MDM2
Gene Name:
MDM2 proto-oncogene
Gene Aliases:
ACTFS, HDMX, hdm2
Chromosome Location:
Chr.12: 68808149 - 68845544 on Build GRCh38
UniGene ID:
Hs.484551
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122297
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145337.2
NP_001138809.1
4
275
NM_001145339.2
NP_001138811.1
4
525
NM_002392.5
NP_002383.2
4
525
XM_005268872.4
XP_005268929.1
4
453
XM_006719399.3
XP_006719462.1
2
108
GenBank mRNA
AF092843.1
-
4
233
AF092844.1
-
4
233
AF385323.1
-
3
201
AJ430612.1
-
3
201
AJ430614.1
-
3
201
AJ491698.1
-
3
201
AJ491699.1
-
4
288
AJ491700.1
-
3
201
AK290341.1
-
4
475
AM902642.1
-
4
294
AM902646.1
-
3
201
AM902647.1
-
4
288
AM902650.1
-
3
201
AM902651.1
-
3
201
AM902654.1
-
3
201
AM902655.1
-
3
201
AM902658.1
-
4
288
AM902659.1
-
3
201
AM902660.1
-
3
201
AM902666.1
-
3
201
AM902667.1
-
3
201
AM902668.1
-
3
201
AM902669.1
-
3
201
AM902670.1
-
2
126
AM902672.1
-
3
201
AM902678.1
-
3, 4
201
AM902680.1
-
3
201
AM902681.1
-
3
201
BC152384.1
-
3
204
BC152390.1
-
3
204
BE296905.1
-
4
490
BT007258.1
-
3
201
EU076747.1
-
4
275
EU076749.1
-
4
275
GQ848196.1
-
2
126
HM213901.1
-
3
201
JX015379.1
-
3
201
JX015390.1
-
3
201
JX015391.1
-
3
201
JX015392.1
-
3
201
M92424.1
-
4
512
U33203.1
-
3
201
Z12020.1
-
4
512
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
blood vessel remodeling
regulation of heart rate
atrioventricular valve morphogenesis
endocardial cushion morphogenesis
ventricular septum development
atrial septum development
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
traversing start control point of mitotic cell cycle
positive regulation of cell proliferation
response to toxic substance
response to carbohydrate
response to iron ion
positive regulation of gene expression
negative regulation of protein processing
viral process
protein ubiquitination
protein sumoylation
peptidyl-lysine modification
protein destabilization
response to magnesium ion
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
regulation of protein catabolic process
response to cocaine
response to drug
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to morphine
negative regulation of DNA damage response, signal transduction by p53 class mediator
establishment of protein localization
response to ether
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
response to antibiotic
positive regulation of protein export from nucleus
response to steroid hormone
cardiac septum morphogenesis
cellular response to hydrogen peroxide
negative regulation of cell cycle arrest
cellular response to antibiotic
cellular response to vitamin B1
cellular response to alkaloid
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to estrogen stimulus
cellular response to hypoxia
cellular response to UV-C
regulation of signal transduction by p53 class mediator
negative regulation of signal transduction by p53 class mediator
p53 binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
SUMO transferase activity
enzyme binding
ubiquitin protein ligase binding
identical protein binding
peroxisome proliferator activated receptor binding
ubiquitin protein ligase activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0