Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002809.3 | NP_002800.2 | 2 | 439 | ||
GenBank mRNA | AK022896.1 | - | 2 | 350 | ||
AK054611.1 | - | 2 | 383 | |||
AK055799.1 | - | 3 | 320 | |||
AK094206.1 | - | 3 | 361 | |||
AK293539.1 | - | 3 | 325 | |||
AK300973.1 | - | 3 | 355 | |||
BC000074.2 | - | 2 | 339 | 3505074 | ||
BC004859.2 | - | 2 | 301 | 3832823 | ||
BC012302.1 | - | 2 | 387 | 3868681 | ||
BC020518.1 | - | 2 | 337 | 3901555 | ||
BC025686.1 | - | 2 | 337 | 5205360 | ||
BT007217.1 | - | 2 | 236 | |||
CR456772.1 | - | 2 | 236 | |||
D67025.1 | - | 2 | 393 | |||
HY034626.1 | - | 2 | 440 |