Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181718.3 | NP_859069.2 | 1 | 798 | ||
XM_017023107.1 | XP_016878596.1 | 2 | 779 | |||
GenBank mRNA | AF070642.1 | - | 1 | 139 | ||
BC009033.1 | - | 1 | 525 | |||
BC029136.1 | - | 1 | 234 | |||
BC034275.3 | - | 1 | 271 | |||
BC040933.2 | - | 1 | 310 | |||
BC085011.1 | - | 1 | 646 | |||
BC126319.1 | - | 1 | 766 | 8992035 | ||
BC126321.1 | - | 1 | 766 | 8992037 | ||
BC144152.1 | - | 1 | 769 | 9052672 |