Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000130.4 | NP_000121.2 | 1, 2 | 297 | ||
GenBank mRNA | AK226079.1 | - | 1, 2 | 215 | ||
AK291613.1 | - | 1, 2 | 247 | |||
AU279657.1 | - | 1, 2 | 297 | |||
M14335.1 | - | 1, 2 | 228 | |||
M16967.1 | - | 1, 2 | 242 |