Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000130.4 | NP_000121.2 | 5 | 780 | ||
XM_017000660.1 | XP_016856149.1 | 4 | 789 | |||
GenBank mRNA | AK226079.1 | - | 5 | 698 | ||
AK291613.1 | - | 5 | 730 | |||
AK300469.1 | - | 4 | 638 | |||
M14335.1 | - | 5 | 711 | |||
M16967.1 | - | 5 | 725 |